missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate, TRC

Product Code. 30006045
Change view
Click to view available options
Quantity:
500 mL
1 L
2 L
Unit Size:
1 L
2 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006045 2 L 2 L
30012824 1 L 1 L
17901685 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006045 Supplier Toronto Research Chemicals Supplier No. TRCE6783502L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4 H8 O2
Molecular Weight (g/mol) 88.11
Synonym Acetic acid ethyl ester, Acetic acid, ester with EtOH (7CI), Ethyl acetate, Acetic acid ethyl ester, Acetic ether, Acetidin, Acetoxyethane, EtOAc, Ethyl ethanoate, NSC 70930, Vinegar naphtha
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Quantity 2 L
InChI Formula InChI=1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3
Formula Weight 88.0524
Chemical Name or Material Ethyl acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.