Learn More
Ethyl 7-chloro-1-(2,4-difluorophenyl)-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 100491-29-0 |
| Molecular Formula | C17H10ClF3N2O3 |
| MDL Number | MFCD01863285 |
| Quantity | 1 g |
| SMILES | O=C(C1=CN(C2=CC=C(F)C=C2F)C3=C(C=C(F)C(Cl)=N3)C1=O)OCC |
| Molecular Weight (g/mol) | 382.72 |
| Percent Purity | 97% |
| Chemical Name or Material | Ethyl 7-chloro-1-(2,4-difluorophenyl)-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate |