Learn More
Ethyl 5-amino-1-benzofuran-2-carboxylate, TRC
Specifications
Specifications
| Chemical Name or Material | Ethyl 5-amino-1-benzofuran-2-carboxylate |
| CAS | 174775-48-5 |
| Molecular Formula | C11H11NO3 |
| Quantity | 10 g |
| Synonym | 5-Aminobenzofuran-2-carboxylic acid ethyl ester, Ethyl 5-aminobenzofuran-2-carboxylate |
| InChI Formula | InChI=1S/C11H11NO3/c1-2-14-11(13)10-6-7-5-8(12)3-4-9(7)15-10/h3-6H,2,12H2,1H3 |
| SMILES | CCOC(=O)c1oc2ccc(N)cc2c1 |
| Molecular Weight (g/mol) | 205.21 |
| Formula Weight | 205.0739 |