Learn More
Ethyl 4-Oxo-3-piperidinecarboxylate Hydrochloride, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 4644-61-5 |
| Molecular Formula | C8H14ClNO3 |
| MDL Number | MFCD00043267 |
| Quantity | 5 g |
| SMILES | O=C(C1CNCCC1=O)OCC.[H]Cl |
| Molecular Weight (g/mol) | 207.66 |
| Percent Purity | 97% |
| Chemical Name or Material | Ethyl 4-Oxo-3-piperidinecarboxylate Hydrochloride |