missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyl (4-formyl-2-methoxyphenoxy)acetate

Product Code. 11325569
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11325569 1 g 1g
11315569 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11325569 Supplier Alfa Aesar™ Supplier No. H50491.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 51264-70-1
Molecular Formula C12H14O5
Molecular Weight (g/mol) 238.24
MDL Number MFCD01764358
InChI Key SVYWLZTVBMJCGG-UHFFFAOYSA-N
Synonym ethyl 2-4-formyl-2-methoxyphenoxy acetate, ethyl 4-formyl-2-methoxyphenoxy acetate, 4-formyl-2-methoxy-phenoxy-acetic acid ethyl ester, acmc-1ap2o, 4-carbethoxymethoxy-3-methoxybenzaldehyde, 2-methoxy-4-formyl-phenoxyessigsaureathylester, 4-formyl-2-methoxyphenoxy acetic acid ethyl ester, acetic acid, 2-4-formyl-2-methoxyphenoxy-, ethyl ester
PubChem CID 968872
IUPAC Name ethyl 2-(4-formyl-2-methoxyphenoxy)acetate
SMILES CCOC(=O)COC1=C(OC)C=C(C=O)C=C1

Specifications

Quantity 1 g
Formula Weight 238.24
Chemical Name or Material Ethyl (4-formyl-2-methoxyphenoxy)acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.