missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-bromophenylacetate, 98%

Product Code. p-7026256
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11449697 5 g 5g
11459697 25 g 25g
11469697 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11449697 Supplier Thermo Scientific Alfa Aesar Supplier No. B22165.06
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14062-25-0 | C10H11BrO2 | 243.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14062-25-0
Molecular Formula C10H11BrO2
Molecular Weight (g/mol) 243.1
MDL Number MFCD00016333
InChI Key ZFDCWHPNBWPPHG-UHFFFAOYSA-N
Synonym ethyl 4-bromophenylacetate, ethyl 2-4-bromophenyl acetate, 4-bromophenylacetic acid ethyl ester, benzeneacetic acid, 4-bromo-, ethyl ester, 4-bromo-phenyl-acetic acid ethyl ester, ethyl 4-bromophenyl acetate, 4-bromo-benzeneacetic acid ethyl ester, ethyl-4-bromophenylacetate, pubchem20039, acmc-209ckw
PubChem CID 7020609
IUPAC Name ethyl 2-(4-bromophenyl)acetate
SMILES CCOC(=O)CC1=CC=C(C=C1)Br

Specifications

Melting Point 29°C to 31°C
Boiling Point 88°C to 90°C (0.35 mmHg)
Quantity 5 g
Formula Weight 243.1
Percent Purity 98%
Chemical Name or Material Ethyl 4-bromophenylacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.