missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Ethyl 3-chloropropionate, 98%
Description
Applications
It can be used to produce (1-ethoxycyclopropyloxy)trimethylsilane by heating.
Solubility
Immiscible in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It can be used to produce (1-ethoxycyclopropyloxy)trimethylsilane by heating.
Solubility
Immiscible in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Boiling Point | 162°C to 164°C |
| Flash Point | 61°C (142°F) |
| Quantity | 100 g |
| Beilstein | 385698 |
| Synonym | ethyl 3-chloropropionate, 3-chloropropionic acid ethyl ester, propanoic acid, 3-chloro-, ethyl ester, ethyl beta-chloropropionate, propionic acid, 3-chloro-, ethyl ester, ethyl .beta.-chloropropionate, 3-chloropropionicacidethylester, 3-chloro-propanoicacidethylester, ethyl-3-chlorpropanoat, ethyl3-chloropropionate |
| Formula Weight | 136.58 |
| Percent Purity | 98% |
| Chemical Name or Material | Ethyl 3-chloropropionate |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?