Learn More
Ethyl 2-thiouracil-5-carboxylate, 99%
Product Code. 11468906
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11468906 5 g 5g
11478906 25 g 25g
2 options
Product Code. 11468906
Brand: Thermo Scientific Alfa Aesar A17046.06
Request Bulk or Custom Format
CAS: 38026-46-9 | C7H8N2O3S | 200.21 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 38026-46-9
Molecular Formula C7H8N2O3S
Molecular Weight (g/mol) 200.21
MDL Number MFCD00006074
InChI Key FQFSHLBWRUOCPX-UHFFFAOYSA-N
Synonym 5-carbethoxy-2-thiouracil, ethyl 2-thiouracil-5-carboxylate, 2-mercapto-5-carbethoxyuracil, 5-ethoxycarbonyl-2-thiouracil, uracil, 5-carbethoxy-2-thio, ethyl 4-hydroxy-2-mercaptopyrimidine-5-carboxylate, 5-carboethoxy-2-thiouracil, ethyl 4-oxo-2-thioxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate, 5-pyrimidinecarboxylic acid, 1,2,3,4-tetrahydro-4-oxo-2-thio-, ethyl ester, 5-pyrimidinecarboxylic acid, 1,2,3,4-tetrahydro-4-oxo-2-thioxo-, ethyl ester
PubChem CID 735952
IUPAC Name ethyl 4-oxo-2-sulfanylidene-1H-pyrimidine-5-carboxylate
SMILES CCOC(=O)C1=CNC(=S)NC1=O
Specifications
Melting Point ∼250°C (decomposition)
Quantity 5 g
Beilstein 190867
Formula Weight 200.22
Percent Purity 99%
Chemical Name or Material Ethyl 2-thiouracil-5-carboxylate