accessibility menu, dialog, popup

UserName

CAS RN 38026-46-9

H3COONHSHNO

CAS RN 38026-46-9

IUPAC Name: ethyl 4-oxo-2-sulfanylidene-1,2,3,4-tetrahydropyrimidine-5-carboxylate
Synonyms: ethyl 4-oxo-2-sulfanylidene-1,3-dihydropyrimidine-5-carboxylate
Molecular Weight (g/mol): 200.21
Molecular Formula: C7H8N2O3S
InChi Key: FQFSHLBWRUOCPX-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CNC(=S)NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results