Learn More
Ethyl 2-methyl-2-(methylthio)propionate, 98%, Thermo Scientific™
Product Code. 11465177
Change view
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11465177 10 g 10g
11455177 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11465177 Supplier Thermo Scientific Alfa Aesar Supplier No. B22597.09
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 49773-24-2
Molecular Formula C7H14O2S
Molecular Weight (g/mol) 162.247
MDL Number MFCD00053099
InChI Key JLURSSXMQSLZQL-UHFFFAOYSA-N
Synonym ethyl 2-methyl-2-methylthio propionate, propanoic acid, 2-methyl-2-methylthio-, ethyl ester, ethyl 2-methyl-2-methylthio propanoate, ethyl 2-methyl-2-methylsulfanyl propanoate, acmc-20al0x, ethyl 2-methylthio isobutyrate, 2-methyl-2-methylthio propionic acid ethyl ester, 2-methyl-2-methylthio-propanoic acid ethyl ester
PubChem CID 3016499
IUPAC Name ethyl 2-methyl-2-methylsulfanylpropanoate
SMILES CCOC(=O)C(C)(C)SC
Specifications
Quantity 10 g
Formula Weight 162.25
Percent Purity 98%
Chemical Name or Material Ethyl 2-methyl-2-(methylthio)propionate