missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-ethoxymethylene-4,4,4-trifluoro-3-oxobutyrate, 97%

Product Code. p-7028547
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11455680 10 g 10g
11465680 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11455680 Supplier Thermo Scientific Alfa Aesar Supplier No. H25927.09
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 571-55-1 | C9H11F3O4 | 240.178 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 571-55-1
Molecular Formula C9H11F3O4
Molecular Weight (g/mol) 240.178
MDL Number MFCD02677683
InChI Key XNGGOXOLHQANRB-AATRIKPKSA-N
Synonym ethyl ethoxymethylene-3-oxo-4,4,4-trifluorobutyrate, ethyl 2-ethoxymethylene-4,4,4-trifluoro-3-oxobutyrate, e-ethyl 2-ethoxymethylene-4,4,4-trifluoro-3-oxobutanoate, ethyl 2-ethoxymethylene-4,4,4-trifluoroacetoacetate, ethyl 2e-2-ethoxymethylidene-4,4,4-trifluoro-3-oxobutanoate, ethyl 2-ethoxymethylidene-4,4,4-trifluoro-3-oxobutanoate, ethyl ethoxymethylene-3-oxo-4,4,4-trifluoro-butyrate, ethyl 2-ethoxymethylene-4,4,4-trifluoro-3-oxobutanoate, ethyl 2e-3-ethoxy-2-2,2,2-trifluoroacetyl prop-2-enoate, 2e-2-ethoxymethylidene-4,4,4-trifluoro-3-oxobutanoic acid ethyl ester
PubChem CID 6185754
IUPAC Name ethyl (2E)-2-(ethoxymethylidene)-4,4,4-trifluoro-3-oxobutanoate
SMILES CCOC=C(C(=O)C(F)(F)F)C(=O)OCC

Specifications

Density 1.235
Boiling Point 80°C to 82°C (1 mmHg)
Flash Point 104°C (220°F)
Refractive Index 1.429
Quantity 10 g
Formula Weight 240.18
Percent Purity 97%
Chemical Name or Material Ethyl 2-ethoxymethylene-4,4,4-trifluoro-3-oxobutyrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.