missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (2-Cyano-2-(2-(3,5-dichloro-4-((4-chlorophenyl)(cyano)methyl)phenyl)hydrazono)acetyl)carbamate-13C3,15N2, TRC

Product Code. 30380199
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380199 25 mg 25mg
30371571 10 mg 10mg
30369063 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380199 Supplier Toronto Research Chemicals Supplier No. TRCE89527225MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl (2-Cyano-2-(2-(3,5-dichloro-4-((4-chlorophenyl)(cyano)methyl)phenyl)hydrazono)acetyl)carbamate-13C3,15N2
Molecular Formula C17 13C3 H14 Cl3 N3 15N2 O3
Quantity 25 mg
InChI Formula InChI=1S/C20H14Cl3N5O3/c1-2-31-20(30)26-19(29)17(10-25)28-27-13-7-15(22)18(16(23)8-13)14(9-24)11-3-5-12(21)6-4-11/h3-8,14,27H,2H2,1H3,(H,26,29,30)/b28-17+/i17+1,19+1,20+1,26+1,28+1
SMILES N#CC(C1=C(Cl)C=C(N/[15N]=[13C]([13C]([15NH][13C](OCC)=O)=O)\C#N)C=C1Cl)C2=CC=C(Cl)C=C2
IUPAC Name ethyl (E)-(2-cyano-2-(2-(3,5-dichloro-4-((4-chlorophenyl)(cyano)methyl)phenyl)hydrazineylidene-1-15N)acetyl-13C2)carbamate-13C-15N
Molecular Weight (g/mol) 483.68
Formula Weight 482.02
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.