missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-chloronicotinate, 98%

Product Code. 11484257
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11484257 50 g 50g
11474257 10 g 10g
11494257 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11484257 Supplier Thermo Scientific Alfa Aesar Supplier No. B20359.18
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1452-94-4 | C8H8ClNO2 | 185.607 g/mol

Ethyl 2-chloronicotinate,is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-chloronicotinate,is used as a pharmaceutical intermediate.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1452-94-4
Molecular Formula C8H8ClNO2
Molecular Weight (g/mol) 185.607
MDL Number MFCD00674129
InChI Key PMIMPBYTPPRBGD-UHFFFAOYSA-N
Synonym ethyl 2-chloronicotinate, 2-chloronicotinic acid ethyl ester, ethyl-2-chloronicotinate, 2-chloro-nicotinic acid ethyl ester, ethyl 2-chloro-3-pyridinecarboxylate, 3-pyridinecarboxylic acid, 2-chloro-, ethyl ester, 2-chloro-3-pyridinecarboxylic acid ethyl ester, ethyl 2chloronicotinate, acmc-1brbe, ethyl 2-chloro-nicotinate
PubChem CID 2796614
IUPAC Name ethyl 2-chloropyridine-3-carboxylate
SMILES CCOC(=O)C1=C(N=CC=C1)Cl

Specifications

Quantity 50 g
Formula Weight 185.61
Percent Purity 98%
Chemical Name or Material Ethyl 2-chloronicotinate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.