missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-chloro-4-methylpyrimidine-5-carboxylate, 95%

Product Code. 15406316
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15406316 5 g 5g
15496306 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15406316 Supplier Thermo Scientific Alfa Aesar Supplier No. H61158.06
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 188781-08-0 | C8H9ClN2O2 | 200.622 g/mol

It is an active pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Store in cool dry ventilated place. Ensure proper ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 188781-08-0
Molecular Formula C8H9ClN2O2
Molecular Weight (g/mol) 200.622
MDL Number MFCD03425963
InChI Key XJNJPQSXRUJCIB-UHFFFAOYSA-N
Synonym 2-chloro-4-methyl-pyrimidine-5-carboxylic acid ethyl ester, 5-pyrimidinecarboxylic acid, 2-chloro-4-methyl-, ethyl ester, 2-chloro-5-ethoxycarbonyl-4-methylpyrimidine, ethyl2-chloro-4-methylpyrimidine-5-carboxylate, 2-chloro-4-methyl-5-pyrimidinecarboxylic acid ethyl ester
PubChem CID 21220624
IUPAC Name ethyl 2-chloro-4-methylpyrimidine-5-carboxylate
SMILES CCOC(=O)C1=CN=C(N=C1C)Cl

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 200.62
Percent Purity 95%
Chemical Name or Material Ethyl 2-chloro-4-methylpyrimidine-5-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.