Learn More
Ethyl 2-bromo-4,4,4-trifluorobutyrate, 97%
Product Code. 15465317
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15465317 250 mg 250mg
15455317 1 g 1g
2 options
Product Code. 15465317
Brand: Thermo Scientific Alfa Aesar undefined
Request Bulk or Custom Format
CAS: 367-33-9 | C6H8BrF3O2 | 249.027 g/mol
Ethyl 2-bromo-4,4,4-trifluorobutyrate is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl 2-bromo-4,4,4-trifluorobutyrate is used as a pharmaceutical intermediate.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 367-33-9
Molecular Formula C6H8BrF3O2
Molecular Weight (g/mol) 249.027
MDL Number MFCD10687235
InChI Key POVYUFTVLGOXEE-UHFFFAOYSA-N
Synonym 2-bromo-4,4,4-trifluorobutyric acid ethyl ester, ethyl 2-bromo-4,4,4-trifluorobutyrate, ethyl 2-bromo-4,4,4-trifluoro-butanoate, butanoic acid,2-bromo-4,4,4-trifluoro-, ethyl ester
PubChem CID 45594306
IUPAC Name ethyl 2-bromo-4,4,4-trifluorobutanoate
SMILES CCOC(=O)C(CC(F)(F)F)Br
Specifications
Boiling Point 67°C (18 mmHg)
Quantity 250 mg
UN Number UN3265
Formula Weight 249.03
Percent Purity 97%
Chemical Name or Material Ethyl 2-bromo-4,4,4-trifluorobutyrate