Learn More
Ethyl 2-Bromo-3-oxobutanoate (>80%), TRC
Specifications
Specifications
| Chemical Name or Material | Ethyl 2-Bromo-3-oxobutanoate |
| CAS | 609-13-2 |
| Molecular Formula | C6H9O3Br |
| Quantity | 1 g |
| Synonym | 2-Bromo-3-oxo-butyric acid ethyl ester, 2-Bromo-3-oxobutanoic acid ethyl ester, Ethyl 2-bromo-3-oxobutanoate, Ethyl 2-bromo-3-oxobutyrate, Ethyl 2-bromoacetoacetate, Ethyl alpha-bromoacetoacetate |
| InChI Formula | InChI=1S/C6H9BrO3/c1-3-10-6(9)5(7)4(2)8/h5H,3H2,1-2H3 |
| SMILES | CCOC(=O)C(Br)C(=O)C |
| IUPAC Name | ethyl 2-bromo-3-oxobutanoate |
| Molecular Weight (g/mol) | 209.03 |
| Formula Weight | 207.9735 |