Learn More
Ethyl 2,4-dichloropyrimidine-5-carboxylate, 98%
Product Code. 15412149
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15412149 1 g 1g
15459146 250 mg 250mg
15422149 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15412149 Supplier Thermo Scientific Alfa Aesar Supplier No. H64788.03
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
CAS: 51940-64-8 | C7H6Cl2N2O2 | 221.037 g/mol
It's employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It′s employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 51940-64-8
Molecular Formula C7H6Cl2N2O2
Molecular Weight (g/mol) 221.037
MDL Number MFCD09910281
InChI Key SRJBDGLSCPDXBL-UHFFFAOYSA-N
PubChem CID 104020
IUPAC Name ethyl 2,4-dichloropyrimidine-5-carboxylate
SMILES CCOC(=O)C1=CN=C(N=C1Cl)Cl
Specifications
Melting Point 36°C to 37°C
Color White
Quantity 1 g
Beilstein 152173
Synonym ethyl 2,4-dichloro-5-pyrimidinecarboxylate, 5-pyrimidinecarboxylic acid, 2,4-dichloro-, ethyl ester, ethyl 2,4-dichloro-pyrimidine-5-carboxylate, 2,4-dichloro-pyrimidine-5-carboxylic acid ethyl ester, 2,4-dichloro-5-ethoxycarbonylpyrimidine, 2,4,-dichloro-5-pyrimidinecarboxylic acid ethyl ester, 2,4-dichloro-5-pyrimidinecarboxylic acid ethyl ester, 2,4-dichloropyrimidine-5-carboxylic acid ethyl ester, pubchem24010, ethyl2,4-dichloro-5-pyrimidinecarboxylate
Solubility Information Slightly soluble in water.
Formula Weight 221.04
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Ethyl 2,4-dichloropyrimidine-5-carboxylate