Learn More
Ethyl 1-naphthoate, 97%
Product Code. 11382037
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11382037 50 g 50g
11372037 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11382037 Supplier Thermo Scientific Alfa Aesar Supplier No. L07760.18
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
CAS: 3007-97-4 | C13H12O2 | 200.237 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3007-97-4
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.237
MDL Number MFCD00016805
InChI Key XCTLDQQOHIEUCJ-UHFFFAOYSA-N
Synonym ethyl 1-naphthoate, ethyl alpha-naphthoate, 1-naphthoic acid, ethyl ester, ethyl alfa-naphthoate, 1-naphthalenecarboxylic acid, ethyl ester, 1-naphthoic acid ethyl ester, naphthalene-1-carboxylic acid ethyl ester, ethyl naphthalenecarboxylate, ethyl a-naphthoate, ethylalpha-naphthoate
PubChem CID 76364
IUPAC Name ethyl naphthalene-1-carboxylate
SMILES CCOC(=O)C1=CC=CC2=CC=CC=C21
Specifications
Density 1.11
Boiling Point 308°C to 310°C
Refractive Index 1.593
Quantity 50 g
Beilstein 2047857
Formula Weight 200.24
Percent Purity 97%
Chemical Name or Material Ethyl 1-naphthoate