Learn More
Ethyl (1,1-Dioxido-3-oxo-1,2-benzisothiazol-2(3H)-yl)acetate, Mikromol™
Product Code. 17990986
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17990986 25 mg 25mg
17972546 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17990986 Supplier Mikromol Supplier No. MM0084.050025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ethyl (1,1-Dioxido-3-oxo-1,2-benzisothiazol-2(3H)-yl)acetate
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Ethyl (1,1-Dioxido-3-oxo-1,2-benzisothiazol-2(3H)-yl)acetate
CAS 24683-20-3
Analyte or Component Names Ethyl (1,1-Dioxido-3-oxo-1,2-benzisothiazol-2(3H)-yl)acetate
Shipping Condition Room Temperature
Molecular Formula C11 H11 N O5 S
Certifications/Compliance ISO 17025
Synonym Ethyl (1,1-Dioxido-3-oxo-1,2-benzisothiazol-2(3H)-yl)acetate, Piroxicam Imp. E (EP), Piroxicam Impurity E
InChI Formula InChI=1S/C11H11NO5S/c1-2-17-10(13)7-12-11(14)8-5-3-4-6-9(8)18(12,15)16/h3-6H,2,7H2,1H3
SMILES CCOC(=O)CN1C(=O)c2ccccc2S1(=O)=O
IUPAC Name ethyl 2-(1,1,3-trioxo-1,2-benzothiazol-2-yl)acetate
Molecular Weight (g/mol) 269.27
Quantity 25 mg
Formula Weight 269.0358 g/mol