missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1,1,2,2-tetrafluoroethyl ether, 97%, Thermo Scientific™

Product Code. 11475347
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475347 5 g 5g
11485347 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.
Product Code. 11475347 Supplier Thermo Scientific Alfa Aesar Supplier No. B22703.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 512-51-6
Molecular Formula C4H6F4O
Molecular Weight (g/mol) 146.09
MDL Number MFCD00069162
InChI Key HBRLMDFVVMYNFH-UHFFFAOYSA-N
Synonym ethyl 1,1,2,2-tetrafluoroethyl ether, 374pce-beta-gamma, 1,1,2,2-tetrafluoroethyl ethyl ether, ethane, 1-ethoxy-1,1,2,2-tetrafluoro, ether, ethyl 1,1,2,2-tetrafluoroethyl, acmc-1anqb, 1,1,2,2-tetrafluoroethyl ether, 1,1,2,2-tetrafluoroethoxyethane, 1,1,2,2-tetrafluoro ethyl ether, 3-oxa-1,1,2,2-tetrafluoropentane
PubChem CID 120272
IUPAC Name 1-ethoxy-1,1,2,2-tetrafluoroethane
SMILES CCOC(F)(F)C(F)F

Specifications

Boiling Point 56°C to 58°C
Flash Point −15°C (−5°F)
Linear Formula HCF2CF2OCH2CH3
Quantity 5 g
UN Number UN3271
Formula Weight 146.08
Percent Purity 97%
Chemical Name or Material Ethyl 1,1,2,2-tetrafluoroethyl ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.