missing translation for 'onlineSavingsMsg'
Learn More

Ethoxyquin, MedChemExpress

Product Code. 16492339
Encompass_Preferred
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 16492339

Brand: Medchem Express HYB1425

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ethoxyquin is an antioxidant which has been used in animal feed for many years and also an inhibitor of heat shock protein 90 (Hsp90).

Ethoxyquin is an antioxidant which has been used in animal feed for many years and also an inhibitor of heat shock protein 90 (Hsp90).

Specifications

Chemical Name or Material Ethoxyquin
CAS 91-53-2
Color Colorless
Purity Grade Notes Research
Molecular Formula C14H19NO
Quantity 1 g
Solubility Information DMSO : ≥ 50 mg/mL (230.09 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CC1=CC(C)(C)NC2=C1C=C(OCC)C=C2
Molecular Weight (g/mol) 217.31
Formula Weight 217.31
Percent Purity 98.29%
Grade Research
Physical Form Liquid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.