Learn More
Ethenesulfonyl chloride, TRC
Product Code. 30372474
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30372474 500 mg 500mg
30371463 250 mg -
30386638 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30372474 Supplier Toronto Research Chemicals Supplier No. TRCE677750500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 6608-47-5
Molecular Formula C2H3ClO2S
Molecular Weight (g/mol) 126.56
Synonym Vinylsulfonyl chloride
SMILES ClS(=O)(=O)C=C
Spezifikation
Quantity 500 mg
InChI Formula InChI=1S/C2H3ClO2S/c1-2-6(3,4)5/h2H,1H2
Chemical Name or Material Ethenesulfonyl chloride