Learn More
Ethanolamine, Supelco™
Product Code. 15893960
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15893960 2.5 L 2.5 L
15846890 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15893960 Supplier Supelco Supplier No. 1008452500
 Restricted Item
Estimated Shipment: 18-11-2026
For analysis EMSURE™
EMSURE grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 141-43-5
Specifications
Melting Point 10.5°C
Density 1.02 g/cm3 at 20°C
Boiling Point 171°C/1013 hPa
MDL Number MFCD00008183
Quantity 2.5 L
Synonym Ethanolamine, 2-Aminoethanol, Monoethanolamine, Ethanolamine
Molecular Weight (g/mol) 61.08
Grade Research