Learn More
Ethanethiol, Sigma-Aldrich™
Specifications
Specifications
| Chemical Name or Material | Ethanethiol |
| CAS | 75-08-1 |
| Melting Point | -148°C |
| Density | 0.84 g/cm³ at 20°C |
| Boiling Point | 35°C (1013 hPa) |
| Flash Point | -45°C (closed cup) |
| Quantity | 250 g |
| Synonym | Ethanethiol, Ethyl mercaptan, Ethanethiol |
| Molecular Weight (g/mol) | 62.13 |
| Grade | Not Available |