Description
- Integrated controller and metering pump simplify system setup
- Control systems are able to accept pH or ORP electrodes for added convenience
- On/off or proportional control modes allow for robust process control
- Deadband is programmable for greater flexibility
- LCD is easy to read
Specifications
Specifications
| Includes | 6 ft. (1.8 m) Clear PVC and PE Tubing (3⁄16 x 1⁄4 in.), Weighted Filter, and 6 ft. (1.8 m) U.S. Standard Power Cord |
| Flow Rate | 0.26 GPH |
| Frequency | 50/60 Hz |
| Maximum Pressure | 217.5 psi |
| Voltage | 230 V |