Learn More
Etaqualone Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Etaqualone Hydrochloride |
| CAS | 97979-65-2 |
| Molecular Formula | C17H17ClN2O |
| Quantity | 100 mg |
| Synonym | 3-(2-Ethylphenyl)-2-methyl-4(3H)-quinazolinone Monohydrochloride, 3-(o-Ethylphenyl)-2-methyl-4(3H)-quinazolinone Hydrochloride, Aolan Hydrochloride, Athinazone Hydrochloride, Ethinazone Hydrochloride, NSC 96163 Hydrochloride |
| InChI Formula | InChI=1S/C17H16N2O.ClH/c1-3-13-8-4-7-11-16(13)19-12(2)18-15-10-6-5-9-14(15)17(19)20;/h4-11H,3H2,1-2H3;1H |
| SMILES | O=C1C2=CC=CC=C2N=C(C)N1C3=C(CC)C=CC=C3.Cl |
| IUPAC Name | 3-(2-ethylphenyl)-2-methylquinazolin-4(3H)-one hydrochloride |
| Molecular Weight (g/mol) | 300.78 |
| Formula Weight | 300.1 |