Learn More
Cole-Parmer™ ESD Safe Plastic Tweezer with Flat, Duckbill Tips
Product Code. 15657418 Shop All Cole Parmer Products
Change view
Click to view available options
Length (Metric):
11.5 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
15657418 11.5 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15657418 Supplier Cole-Parmer™ Supplier No. 0738717
  • ONLINE EXCLUSIVE PRICE
    24.20 EUR
    15.39 EUR / Pack of 1
    Save 8.81 EUR
    36% Off
Estimated Shipment: 15-12-2026
ESD-safe tweezers, reinforced with carbon fiber, offer excellent wear and abrasion resistance. These tweezers are ideal for handling sensitive materials such as electronic components, micro-mechanical parts, glass, and ceramic substrates.
  • ESD-safe
  • Reinforced with 30% carbon fiber
  • Wear and abrasion resistant
  • Rigind, high tensile and flexural strength
  • Flat, duckbill tips
  • Handle sensitive electronic components safely
Specifications
Length (Metric) 11.5 cm
Material ESD-safe plastic and carbon fiber
Description ESD-safe plastic tweezers
For Use With (Application) Sensitive materials such as electronic components, micro-mechanical parts, glass, and ceramic substrates
Sterility Non-sterile
Tip Style Flat, Duckbill Tips