Learn More
mb2 Swivel Antistatic Work Chair in Polyurethane - Asyncronous - Glides
Product Code. 13416339
Change view
Click to view available options
Color:
Black
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
13416339 Black Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13416339 Supplier mb2 Supplier No. 245B3C1D2E3
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
Swivel antistatic work chair in polyurethane, Asynchronous mechanism, high gas lift with footrest and glides
  • Antistatic work chair
  • Seat and backrest in multilayers of beech and poplar wood, entirely foamed with black antistatic polyurethane
  • Backrest mechanism in height, depth and permanent contact adjustable and in every position lockable
  • Gas height adjustment
  • Antistatic glides
  • 5-spoke base in die-cast polished aluminium
Specifications
Product Type ESD Chair
Color Black
Adjustable (Height) Yes
Adjustable Seat Height, Seat Tilt, Backrest Height, Backrest Tilt
Back Style Ergonomic
Height (Metric) Seat 540 to 780 mm
Includes Five-star Base, Conductive Pads
Quantity 1 Ea.
Seat Material Polyurethane foam
Seating Height 560 to 800 mm
Weight (Metric) 14.30 kg
With Armrest No
With Backrest Yes
With Casters No
Footrest Yes
With Lumbar Support No
With Tilt Yes
Material Polyurethane
Seating Type Stool with Back