Learn More
Reitenspiess™ Erlenmeyer Piston and Bottles Brush
Product Code. 10179460
Change view
Click to view available options
Length (Metric):
340 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10179460 340 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10179460 Supplier Reitenspiess™ Supplier No. 10000434
Estimated Shipment: 19-10-2026
Add to basket
Add to basket
Use Reitenspiess™ Erlenmeyer Piston and Bottles Brush to clean, disinfect and sterilize Ehrlenmeyer laboratory glassware.
  • 45mm head size
  • For Ehrlemnmeyer laboratory glassware
  • Sterile
Specifications
Product Type Erlenmeyer Piston and Bottles Brush
Diameter (Metric) Bristle 45 mm
Length (Metric) 340 mm
Quantity 1 Ea.