Description
- Contamination-free transfer of trays to stable working box
- System optimized for use with multichannel pipettes
- Color-coded trays for simple identification of tips and pipette
- Tips can be attached to pipette from refill trays
- Boxes and trays are autoclavable at 121°C for subsequent use
- epT.I.P.S. Box 2.0 and epT.I.P.S. Set available in Eppendorf Quality
- ACT™-labeled by non-profit My Green Lab™ to help you make an informed product choice when it comes to sustainability
Specifications
Specifications
| Volume (Metric) | 50 to 1000 μL |
| Filtered | Non-filtered |
| Sterility | Non-sterile |
| Color | Colorless |
| Autoclavable | Autoclavable |
| Model | epTIPS |
| Length (Metric) | 71 mm |
| Certifications/Compliance | EN ISO 8655, DIN EN ISO 11607-1 and 2, DIN EN 868-2 to 10 |
| Clean Claims | Eppendorf Quality (Non-sterile) |
| For Use With (Application) | Pipetting liquids,Dispensing liquids,Mixing liquids,Filling,plates and reaction vessels,Filling gels for electrophoresis,Phase extraction and removal of supernatants |
| Show More |