Learn More
Eppendorf™ epT.I.P.S.™ Box 2.0
Product Code. 17801467 Shop All Eppendorf Products
Change view
Click to view available options
Size:
10 to 300 μL
1000 to 1250 μL
1250 μL to 5 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17801467 1000 to 1250 μL Each
17891457 10 to 300 μL Each
17811467 1250 μL to 5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17801467 Supplier Eppendorf™ Supplier No. 0030076257
Save Up to 60% on Liquid Handling
Use promo code 28028
View all promotions
In Stock
Add to basket
Add to basket
Bulk Quote Request
Eppendorf epT.I.P.S.™ Box 2.0, available with 'Eppendorf Quality' pipette tips or as an empty box, improves lab efficiency with innovative TwinLid™ technology, enhanced stackability, and superior ergonomics for safe, organized pipetting.
  • Contamination-free transfer of trays to stable working box
  • System optimized for use with multichannel pipettes
  • Color-coded trays for simple identification of tips and pipette
  • Tips can be attached to pipette from refill trays
  • Boxes and trays are autoclavable at 121°C for subsequent use
  • epT.I.P.S. Box 2.0 and epT.I.P.S. Set available in Eppendorf Quality
  • ACT™-labeled by non-profit My Green Lab™ to help you make an informed product choice when it comes to sustainability
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Color White, Blue
Holds Pipette Tips
Material Polypropylene
Positions Multiple
Shape Square
Stackable Yes
Sterility Non-sterile
For Use With (Equipment) epT.I.P.S reloads
Packaging 1 Empty Reusable Box
Quantity 1 Ea.
Size 1000 to 1250 μL
Includes Box Only
Product Type Empty Tip Box
Array None