Learn More
Eppendorf™ Xplorer™ Electronic Pipette, Single Channel
GREENER_CHOICE
Product Code. p-4389807 Shop All Eppendorf Products
Change view
Click to view available options
Volume (Metric):
0.1 to 2.5 mL
0.2 to 5 mL
0.5 to 10 μL
0.5 to 10 mL
1 to 20 μL
10 to 200 μL
15 to 300 μL
5 to 100 μL
50 to 1000 μL
Color:
Blue
Light Gray
Medium Gray
Orange
Red
Turquoise
Violet
Yellow
Packungsgröße:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Volume (Metric) Color unitSize
10584585 0.5 to 10 μL Medium Gray Each
16281682 1 to 20 μL Light Gray Each
10237554 5 to 100 μL Yellow Each
16222401 10 to 200 μL Yellow Each
10461255 15 to 300 μL Orange Each
10126314 50 to 1000 μL Blue Each
16232401 0.1 to 2.5 mL Red Each
10034384 0.2 to 5 mL Violet Each
10115934 0.5 to 10 mL Turquoise Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 10584585 Lieferant Eppendorf™ Lieferanten-Nr. 4861000015
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Eliminate potential error risks associated with manual pipetting with these pipettes. Eppendorf™ Xplorer™ Electronic Pipettes provide precisely adjustable parameters, reproducible and accurate results. Experience fatigue-free pipetting and maintain full control over pipetting procedures.
  • Electronic single channel pipettes designed to aspirate and dispense liquid volumes from 0.5 μL to 10 mL
  • Intuitive operating concept and ergonomic design
  • Nine different language settings: Chinese, Dutch, English, French, German, Italian, Japanese, Portuguese and Spanish
  • Multifunctional rocker
  • Selection dial
  • Innovative ejector
  • Ergonomic display angle
  • Convenient handle
  • Powerful rechargeable battery
  • Flexible charging concept
  • Spring-loaded tip cone (except 5 and 10 mL)
  • Adjustment to external conditions (geographic height, liquids with high density or similar factors)
TRUSTED_SUSTAINABILITY
Spezifikation
Accuracy ±2.5% at ±0.025μL/1μL, ±1.5% at ±0.075μL/5μL, ±1% at ±0.1μ
Battery Type Lithium-ion
Compatible Tips 20μL epT.I.P.S™ pipette tip
Color Medium Gray
For Use With epT.I.P.S. Pipette Tip (20 μL)
Number of Channels 1
Product Line Xplorer
Type Electronic Pipette
Volume (Metric) 0.5 to 10 μL
Finger Style Ergonomic
For Use With (Equipment) 20μL epT.I.P.S™ pipette tip
Includes Multifunction rocker, spring loaded tip cone, battery and charger
Increments 0.01μL
Multilingual User Interface Yes
Number of Speeds 8
Temperature (English) Operating 41°F to 104°F
Temperature (Metric) Operating 5°C to 40°C
Mehr anzeigen Weniger anzeigen