Learn More
Eppendorf™ ThermoTop™
Product Code. 13527630 Shop All Eppendorf Products
Change view
Click to view available options
Type:
Tube Lid
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
13527630 Tube Lid Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13527630 Supplier Eppendorf™ Supplier No. 5308000003
  • ONLINE EXCLUSIVE PRICE
    610.00 EUR
    566.00 EUR / Pack of 1
    Save 44.00 EUR
    7% Off
View all promotions
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
Use this lid with ThermoMixer™ C/ ThermoStat™ C with SmartBlock™ plates and lab vessels up to 2 mL and ThermoMixer F 0.5/F1.5/F2.0/FP. The Eppendorf ThermoTop™ with condens.protect™ technology heated lid prevents condensation on tube lid and tube wall.
  • Efficient and reliable prevention of condensation on tube lid and wall
  • Get constant reaction conditions for optimal enzymatic performance within your samples
  • Wireless connection with automatic recognition and operation for user-friendly handling
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 2 mL
For Use With (Equipment) Thermomixer F1.5, FP and C, ThermoStat C with Eppendorf SmartBlocks 0.5, 1.5 and 2mL, Plates, PCR 96 and PCR 384
Type Tube Lid
For Use With (Application) Plates, lab vessels up to 2 mL