Learn More
Eppendorf™ Research™ plus, Mechanical Single-Channel Pipettes with Fixed Volume
Product Code. 15763469 Shop All Eppendorf Products
Change view
Click to view available options
Volume (Metric):
10 μL
20 μL
25 μL
50 μL
100 μL
200 μL
250 μL
500 μL
1000 μL
Accuracy:
±0.6%
±0.7%
±0.8%
±1%
±1.2%
Color:
Blue
Gray
Yellow
Unit Size:
Each
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Accuracy Color Unit Size
15763469 1000 μL ±0.6% Blue Each
15753459 10 μL ±1.2% Gray Each
15763459 10 μL ±1.2% Yellow Each
15773459 20 μL ±0.8% Gray Each
15783459 20 μL ±1% Yellow Each
15793459 25 μL ±1% Yellow Each
15703469 50 μL ±0.7% Yellow Each
15713469 100 μL ±0.6% Yellow Each
15733469 200 μL ±0.6% Blue Each
15723469 200 μL ±0.6% Yellow Each
15743469 250 μL ±0.6% Blue Each
15753469 500 μL ±0.6% Blue Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 15763469 Supplier Eppendorf™ Supplier No. 3124000121
  • ONLINE EXCLUSIVE PRICE
    213.00 EUR
    198.00 EUR / Pack of 1
    Save 15.00 EUR
    7% Off
View all promotions
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
Bulk Quote Request
Resistant to chemical, thermal, and mechanical stress
  • Control button and ejector's low operating force are positioned for ergonomics
  • 4-digit, magnifying-shape volume display Calibration display and adjustment opening for specific liquids and volumes Ultra light PerfectPiston™ piston system constructed of chemically resistant Fortron™ Quick connection clip allows removal of lower part in two quick steps (not in 10μL pipetters) Spring loaded tip cone promotes tight fit to the tip Color indicates pipetter's volume
TRUSTED_SUSTAINABILITY
Specifications
Accuracy ±0.6%
Autoclavable Autoclavable
Color Blue
For Use With 1000 μL Pipette Tip
Number of Channels 1
Type Fixed Volume Pipette
Volume (Metric) 1000 μL
For Use With (Equipment) 1000μL Pipette tip
Includes Spring loaded tip cone, ejector, blue operating button