Learn More
Epoxystyrene, Sigma-Aldrich™
Specifications
Specifications
| CAS | 96-09-3 |
| Melting Point | -37°C |
| Density | 1.05 g/cm3 at 20°C |
| Boiling Point | 193°C to 195°C/1013 hPa |
| MDL Number | MFCD00005121 |
| Quantity | 250 mL |
| Synonym | Epoxystyrene, Styrene oxide, Phenyloxirane, Phenylethylene oxide, Styrene epoxide, Epoxystyrene |
| Molecular Weight (g/mol) | 120.15 |
| Grade | Research |