Learn More
Epredia™ Epoxy Diagnostic Slides
Product Code. 16389485 Shop All Epredia Products
Change view
Click to view available options
Well Count:
1
3
4
5
8
10
12
Color:
Black
Blue
White
Diameter (Metric):
10 mm Well
11.43 mm Well
13 mm Well
14 mm Well
5 mm Well
6 mm Well
6.7 mm Well
8 mm Well
Unit Size:
Pack of 100
Pack of 1000
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Well Count Color Diameter (Metric) Unit Size
16389485 5 White 8 mm Well Pack of 100
10106260 3 Black 10 mm Well Pack of 100
10588681 10 Black 6.7 mm Well Pack of 100
10777371 8 Black 6 mm Well Pack of 100
13535770 1 White 13 mm Well Pack of 100
10393881 3 Black 14 mm Well Pack of 100
10484681 1 Blue 8 mm Well Pack of 100
12675187 4 Blue 8 mm Well Pack of 100
15283219 4 Blue 8 mm Well Pack of 1000
15391361 4 White 11.43 mm Well Pack of 100
10529261 12 White 5 mm Well Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 16389485 Supplier Epredia™ Supplier No. X1XER307W
  • ONLINE EXCLUSIVE PRICE
    79.15 EUR
    70.27 EUR / Pack of 100
    Save 8.88 EUR
    11% Off
 Restricted Item
Estimated Shipment: 01-02-2027
These hydrophobic coatings bind liquids applied to the wells and keeps fluids in the well area. Clean, flat and suitable for immediate use
  • Binds and keeps liquids applied to the well area
  • Provides wettable wells for staining and growing cell cultures
  • Prevents cross-contamination
  • Hydrophobic coatings are very durable and withstand the strongest stains
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) 25 mm
Width (Metric) 75 mm
Material Epoxy
Thickness (Metric) 1 mm
Type Diagnostic Slide
Well Count 5
Color White
Diameter (Metric) 8 mm Well