Learn More
Gibco™ EpiLife™ Defined Growth Supplement (EDGS)
missing translation for 'orderingAttributeHoverText'
Quantity:
5 mL
missing translation for 'unitSize'
Each
Descripción
A defined, sterile, concentrated (100X) solution intended for use with EpiLife™ medium to culture human epidermal keratinocytes. EDGS is not intended for use with Medium 154. EDGS contains all of the growth factors and hormones necessary for the growth of human epidermal keratinocytes. Each bottle contains the correct amount of supplement for a 500 ml bottle of EpiLife™ medium. EDGS contains:
purified bovine serum albumin
purified bovine transferrin
hydrocortisone
recombinant human insulin-like growth factor type-1 (rhIGF-1)
prostaglandin E2 (PGE2)
recombinant human epidermal growth factor (rhEGF) The bovine products in EDGS have been isolated from animals of North American origin. The isolation process for these components includes steps that inactivate viruses. All components of EDGS are greater than 95% pure.
Especificaciones
Especificaciones
| Content And Storage | minus 20 degrees C |
| Cell Type | Keratinocyte Cells |
| Concentration | 100 X |
| Product Type | Growth Supplement |
| Sterility | Sterile |
| Product Line | Cascade Biologics™, EpiLife™ |
| Quantity | 5 mL |
| Shipping Condition | Dry Ice |
| Species | Human |
For Research Use Only. Not for use in diagnostic procedures.
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido