Learn More
(-)-Epigallocatechin, TRC
Product Code. 17947454
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17947454 1 mg 1mg
17948024 5 mg 5mg
17910005 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17947454 Supplier Toronto Research Chemicals Supplier No. TRCE5884901MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 970-74-1
Molecular Formula C15 H14 O7
Molecular Weight (g/mol) 306.27
Synonym 2H-1-Benzopyran-3,5,7-triol, 3,4-dihydro-2-(3,4,5-trihydroxyphenyl)-, (2R-cis)-, Epigallocatechol (8CI), Gallocatechol, l- (6CI), (2R,3R)-3,4-Dihydro-2-(3,4,5-trihydroxyphenyl)-2H-1-benzopyran-3,5,7-triol, (-)-3,3',4',5,5',7-Flavanhexol, (-)-Epigallocatechin, (-)-Epigallocatechol, (-)-epi-Gallocatechin, (2R,3R)-2-(3,4,5-Trihydroxyphenyl)-3,5,7-trihydroxychroman, 1-epi-3',4',5',5,7-Pentahydroxy-3-flavanol, 3,3',4',5,5',7-Flavanhexol, Antiscurvy factor C2, EGC, Epigallocatechin, Factor C2 (antiscurvy), Galloepicatechin, L-Epigallocatechin, NSC 674039, Sunphenon EGC, epi-Gallocatechin, l-Epigallocatechin, l-Epigallocatechol
IUPAC Name (2R,3R)-2-(3,4,5-trihydroxyphenyl)-3,4-dihydro-2H-chromene-3,5,7-triol
SMILES O[C@@H]1Cc2c(O)cc(O)cc2O[C@@H]1c3cc(O)c(O)c(O)c3
Specifications
Quantity 1 mg
InChI Formula InChI=1S/C15H14O7/c16-7-3-9(17)8-5-12(20)15(22-13(8)4-7)6-1-10(18)14(21)11(19)2-6/h1-4,12,15-21H,5H2/t12-,15-/m1/s1
Formula Weight 306.074
Percent Purity >95%
Chemical Name or Material (-)-Epigallocatechin
Grade HPLC