Learn More
Epigallocatechin 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30318598
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30318598 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30318598 Supplier Dr. Ehrenstorfer Supplier No. DREA13176400AL1000
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (-)-Epigallocatechin
CAS 970-74-1
Analyte or Component Names (-)-Epigallocatechin
Shipping Condition Room Temperature
Molecular Formula C15 H14 O7
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/C15H14O7/c16-7-3-9(17)8-5-12(20)15(22-13(8)4-7)6-1-10(18)14(21)11(19)2-6/h1-4,12,15-21H,5H2/t12-,15-/m1/s1
SMILES O[C@@H]1Cc2c(O)cc(O)cc2O[C@@H]1c3cc(O)c(O)c(O)c3
IUPAC Name (2R,3R)-2-(3,4,5-trihydroxyphenyl)-3,4-dihydro-2H-chromene-3,5,7-triol
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 306.27
Quantity 1 mL
Formula Weight 306.074 g/mol