Learn More
Epichlorohydrin, Sigma-Aldrich™
Product Code. 15869000
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Unit Size:
1 L
100mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15869000 1 L 1 L
15849000 100 mL 100mL
15859000 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15869000 Supplier Sigma Aldrich Supplier No. 8032961000
 Restricted Item
Estimated Shipment: 19-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 106-89-8
Specifications
Melting Point -57°C
Density 1.18 g/cm3 at 20°C
MDL Number MFCD00005132
Quantity 1 L
Synonym Epichlorohydrin, 1-Chloro-2,3-epoxypropane, 2-Chloromethyl oxirane, 2,3-Epoxypropyl chloride, Epichlorohydrin
Molecular Weight (g/mol) 92.52
Grade Research