missing translation for 'onlineSavingsMsg'
Learn More

Epichlorohydrin, >99%, MP Biomedicals™

Product Code. 11466842 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11466842 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11466842 Supplier MP Biomedicals Supplier No. 0215106380
  • ONLINE EXCLUSIVE PRICE
    62.85 EUR
    59.85 EUR / 100g
    Save 3.00 EUR
    4% Off
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 106-89-8
Color Colorless
Density 1.181g/cm3 at 20°C
Assay Percent Range >99%
Molecular Formula C3H5ClO
Quantity 100 g
Synonym epichlorohydrin, 2-chloromethyl oxirane, epichlorhydrin, 1-chloro-2,3-epoxypropane, glycidyl chloride, oxirane, chloromethyl, epichlorhydrine, 1,2-epoxy-3-chloropropane, 2,3-epoxypropyl chloride, chloropropylene oxide
InChI Key BRLQWZUYTZBJKN-UHFFFAOYSA-N
SMILES C1C(O1)CCl
IUPAC Name 2-(chloromethyl)oxirane
Molecular Weight (g/mol) 92.522
PubChem CID 7835
ChEBI CHEBI:37144
Formula Weight 92.52
Physical Form Liquid
Chemical Name or Material Epichlorohydrin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.