Learn More
Epicamphor, TRC
Specifications
Specifications
| Chemical Name or Material | Epicamphor |
| CAS | 10292-98-5 |
| Purity Grade Notes | GC |
| Molecular Formula | C10 H16 O |
| Quantity | 5 mg |
| Synonym | Epicamphor |
| InChI Formula | InChI=1S/C10H16O/c1-9(2)7-4-5-10(9,3)6-8(7)11/h7H,4-6H2,1-3H3/t7-,10+/m1/s1 |
| SMILES | C[C@]12CC[C@H](C(=O)C1)C2(C)C |
| IUPAC Name | (1S,4S)-4,7,7-trimethylbicyclo[2.2.1]heptan-2-one |
| Molecular Weight (g/mol) | 152.233 |
| Show More |