missing translation for 'onlineSavingsMsg'
Learn More

Ephrin-A4 Antibody (MM0263-9H13), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18009387 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18009387 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18009387 Supplier Novus Biologicals Supplier No. NBP212280AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Ephrin-A4 Monoclonal specifically detects Ephrin-A4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ephrin-A4
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0263-9H13
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias EFL4, EPH-related receptor tyrosine kinase ligand 4, ephrin-A4, EPLG4MGC125826, LERK-4, LERK4FLJ57652, ligand of eph-related kinase 4
Gene Symbols EFNA4
Host Species Mouse
Immunogen Human recombinant Ephrin-A4 (extracellular domain)
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 1945
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.