Learn More
Velp Scientifica™ EPDM Tube
Product Code. 12885513 Shop All Velp Products
Change view
Click to view available options
Length (Metric):
11.2 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
12885513 11.2 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12885513 Supplier Velp Scientifica™ Supplier No. 10002412
Estimated Shipment: 21-12-2026
Velp Scientifica™ EPDM Tube for use with Hydrolysis Unit.
  • Spare tubing for Hydrolysis Unit
Specifications
For Use With (Equipment) HU6 Hydrolysis Unit
Product Type Tube
Material EPDM
Quantity 1 Ea.
Diameter (Metric) 6.4 mm Diameter
Length (Metric) 11.2 mm