missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epcam / CD326 Mouse, Clone: rVU-1D9, enQuireBio™
Mouse Monoclonal Antibody
199.00 EUR - 478.00 EUR
Specifications
| Antigen | Epcam / CD326 |
|---|---|
| Clone | rVU-1D9 |
| Concentration | 200μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15950428
|
EnquireBio
4072-MSM20-P0 |
20 μg | |||||||
|
15960428
|
EnquireBio
4072-MSM20-P1 |
100 μg |
554.40 EUR 478.00 EUR / 100µg Save 76.40 EUR 13% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| Epcam / CD326 | |
| 200μg/mL | |
| Monoclonal | |
| Mouse | |
| 10mM PBS with or without 0.05% BSA and 0.05% azide. | |
| 4072 | |
| Small cell lung carcinoma cells | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 40-43kDa |
| rVU-1D9 | |
| Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| NP_002345 | |
| EPCAM | |
| IgG1 κ | |
| Protein A/G Purified | |
| Cell surface | |
| HT29 cells or breast tumor |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title