missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epcam / CD326 Mouse, Clone: rVU-1D9, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 4072-MSM20-P0
This item is not returnable.
View return policy
Specifications
| Epcam / CD326 | |
| Cell surface | |
| rVU-1D9 | |
| Unconjugated | |
| NP_002345 | |
| Mouse | |
| 40-43kDa | |
| Protein A/G Purified | |
| RUO | |
| 4072 | |
| IgG1 κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with or without 0.05% BSA and 0.05% azide. | |
| EPCAM | |
| Small cell lung carcinoma cells | |
| HT29 cells or breast tumor | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction