Learn More
Entacapone, MedChemExpress
Product Code. 16486018
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16486018 10 mg 10mg
16402167 50 mg 50mg
16406436 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16486018 Supplier Medchem Express Supplier No. HY14280
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
MedChemExpress Entacapone is a potent, reversible, peripherally acting and orally active catechol-O-methyltransferase (COMT) inhibitor.
Entacapone inhibits COMT from rat brain, erythrocytes and liver with IC50 values of 10 nM, 20 nM, and 160 nM, respectively. Entacapone is selective for COMT over other catecholamine metabolizing enzymes, including MAO-A, MAO-B, phenolsulphotransferase M (PST-M) and PST-P (IC50s>50 μM). Entacapone can be used for the research of Parkinson's disease. Entacapone serves as a inhibitor of FTO demethylation with an IC50 of 3.5 μM, can be used for the research of metabolic disorders.
Specifications
Chemical Name or Material Entacapone
CAS 130929-57-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C14H15N3O5
Quantity 10 mg
Solubility Information DMSO : 33.33 mg/mL (109.17 mM; Need ultrasonic) ∣H2O : 2 mg/mL (6.55 mM; ultrasonic and adjust pH to 10 with NaOH)
SMILES O=C(N(CC)CC)/C(C#N)=C/C1=CC([N+]([O-])=O)=C(O)C(O)=C1
Molecular Weight (g/mol) 305.29
Formula Weight 305.29
Percent Purity 98.79%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neurodegeneration