missing translation for 'onlineSavingsMsg'
Learn More

ent NAP 226-90, TRC

Product Code. 30179751
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179751 10 mg 10mg
30179736 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30179751 Supplier Toronto Research Chemicals Supplier No. TRCN32500510MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 851086-95-8
Molecular Formula C10 H15 N O
Molecular Weight (g/mol) 165.23
Synonym Phenol, 3-[(1R)-1-(dimethylamino)ethyl]-, 3-[(1R)-1-(Dimethylamino)ethyl]phenol, (R)-3-[1-(Dimethylamino)ethyl]phenol
IUPAC Name 3-[(1R)-1-(dimethylamino)ethyl]phenol
SMILES C[C@@H](N(C)C)c1cccc(O)c1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C10H15NO/c1-8(11(2)3)9-5-4-6-10(12)7-9/h4-8,12H,1-3H3/t8-/m1/s1
Formula Weight 165.1154
Chemical Name or Material ent NAP 226-90
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.