Learn More
Enrofloxacin, Thermo Scientific™
A plant fungicide and antibacterial agent
84.90 EUR - 286.00 EUR
Chemical Identifiers
| CAS | 93106-60-6 |
|---|---|
| Molecular Formula | C19H22FN3O3 |
| Molecular Weight (g/mol) | 359.40 |
| MDL Number | MFCD00792463 |
| InChI Key | SPFYMRJSYKOXGV-UHFFFAOYSA-N |
| Synonym | 1-Cyclopropyl-7-(4-ethyl-1-piperazinyl)-6-fluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic acid |
| PubChem CID | 71188 |
| ChEBI | CHEBI:35720 |
| IUPAC Name | 1-cyclopropyl-7-(4-ethylpiperazin-1-yl)-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid |
| SMILES | CCN1CCN(CC1)C1=C(F)C=C2C(=O)C(=CN(C3CC3)C2=C1)C(O)=O |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15442869
|
Thermo Scientific Alfa Aesar
J60023.06 |
5g |
84.90 EUR
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15452869
|
Thermo Scientific Alfa Aesar
J60023.14 |
25g |
286.00 EUR
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsEnrofloxacin is a broad spectrum antibiotic bactericidal agent used in veterinary medicine to treat animals afflicted with certain bacterial infections.
Solubility
Soluble in chloroform. Slightly soluble in water. Also soluble in dilute KOH
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Chemical Identifiers
| 93106-60-6 | |
| 359.40 | |
| SPFYMRJSYKOXGV-UHFFFAOYSA-N | |
| 71188 | |
| 1-cyclopropyl-7-(4-ethylpiperazin-1-yl)-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid |
| C19H22FN3O3 | |
| MFCD00792463 | |
| 1-Cyclopropyl-7-(4-ethyl-1-piperazinyl)-6-fluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic acid | |
| CHEBI:35720 | |
| CCN1CCN(CC1)C1=C(F)C=C2C(=O)C(=CN(C3CC3)C2=C1)C(O)=O |
Specifications
| 93106-60-6 | |
| C19H22FN3O3 | |
| 5307824 | |
| 1-Cyclopropyl-7-(4-ethyl-1-piperazinyl)-6-fluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic acid | |
| SPFYMRJSYKOXGV-UHFFFAOYSA-N | |
| 1-cyclopropyl-7-(4-ethylpiperazin-1-yl)-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid | |
| 71188 | |
| 359.39 |
| White to Yellow | |
| MFCD00792463 | |
| 14,3592 | |
| Soluble in chloroform; Slightly soluble in water; Also soluble in dilute KOH | |
| CCN1CCN(CC1)C1=C(F)C=C2C(=O)C(=CN(C3CC3)C2=C1)C(O)=O | |
| 359.40 | |
| CHEBI:35720 | |
| Enrofloxacin |
Safety and Handling
missing translation for 'rtecsNumber' : VB1993650
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.