missing translation for 'onlineSavingsMsg'
Learn More

Enrofloxacin, Thermo Scientific™

A plant fungicide and antibacterial agent

Brand:  Thermo Scientific Alfa Aesar J60023.14

Quantity 25g

 View more versions of this product

Product Code. 15452869

  • 286.00 EUR / 25g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Enrofloxacin is a broad spectrum antibiotic bactericidal agent used in veterinary medicine to treat animals afflicted with certain bacterial infections.

Solubility
Soluble in chloroform. Slightly soluble in water. Also soluble in dilute KOH

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

93106-60-6
359.40
SPFYMRJSYKOXGV-UHFFFAOYSA-N
71188
1-cyclopropyl-7-(4-ethylpiperazin-1-yl)-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
C19H22FN3O3
MFCD00792463
1-Cyclopropyl-7-(4-ethyl-1-piperazinyl)-6-fluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic acid
CHEBI:35720
CCN1CCN(CC1)C1=C(F)C=C2C(=O)C(=CN(C3CC3)C2=C1)C(O)=O

Specifications

93106-60-6
25g
MFCD00792463
14,3592
Soluble in chloroform; Slightly soluble in water; Also soluble in dilute KOH
CCN1CCN(CC1)C1=C(F)C=C2C(=O)C(=CN(C3CC3)C2=C1)C(O)=O
359.40
CHEBI:35720
Enrofloxacin
White to Yellow
C19H22FN3O3
5307824
1-Cyclopropyl-7-(4-ethyl-1-piperazinyl)-6-fluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic acid
SPFYMRJSYKOXGV-UHFFFAOYSA-N
1-cyclopropyl-7-(4-ethylpiperazin-1-yl)-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
71188
359.39
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.